6-Amino-4-hydroxyquinoline 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyquinolin-6-amine, 6-Aminoquinolin-4-ol
OR60187-250MG
APOSOR60187-250MGEA
763
EUR
APOSOR60187-250MG
6-Amino-4-hydroxyquinoline 95%
6-Amino-4-hydroxyquinoline
Formel:
C₉H₈N₂O Molecular Weight: 160,173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261102 CAS-Nummer: 56717-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...