8-Bromo-4-hydroxyquinoline 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Bromoquinoline-4-ol

OR60188-1G OR60188-5G OR60188-25G
APOSOR60188-1GEA 43.5 EUR
APOSOR60188-1G APOSOR60188-5G APOSOR60188-25G
8-Bromo-4-hydroxyquinoline 95+%
8-Bromo-4-hydroxyquinoline
Formel: C₉H₆BrNO
Molecular Weight: 224.054 g/mol
Schmelzpunkt: 168…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272389
CAS-Nummer: 57798-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR