8-Bromo-5-chloroquinoline

Lieferant: Apollo Scientific
Achtung

OR60189-1G
APOSOR60189-1GEA 102 EUR
APOSOR60189-1G
8-Bromo-5-chloroquinoline
8-Bromo-5-chloroquinoline
Formel: C₉H₅BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD13193679
CAS-Nummer: 1154741-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR