1-Phenylcyclopropanamine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR60203-5G
APOSOR60203-5GEA 161 EUR
APOSOR60203-5G
1-Phenylcyclopropanamine hydrochloride 95%
1-Phenylcyclopropanamine hydrochloride
Formel: C₉H₁₂ClN
Molecular Weight: 169,6500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07995722
CAS-Nummer: 73930-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR