4-Ethinylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Pyridylacetylene
OR60207-1G
APOSOR60207-1GEA
62.5
EUR
APOSOR60207-1G
4-Ethinylpyridin
4-Ethinylpyridin
Formel:
C₇H₅N Molecular Weight: 103,12 g/mol Schmelzpunkt: 98 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD04038420 CAS-Nummer: 2510-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...