Imidazo[1,2-a]pyridine-6-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR60212-1G
OR60212-250MG
APOSOR60212-1GEA
338
EUR
APOSOR60212-1G
APOSOR60212-250MG
Imidazo[1,2-a]pyridine-6-carboxamide
Imidazo[1,2-a]pyridine-6-carboxamide
Formel:
C₈H₇N₃O Schmelzpunkt: 202…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13176747 CAS-Nummer: 103313-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...