10-Brom-1-decen 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR60224-25G
APOSOR60224-25GEA
92.8
EUR
APOSOR60224-25G
10-Brom-1-decen 95%
10-Bromodec-1-ene
Formel:
C₁₀H₁₉Br Molecular Weight: 219,1600 g/mol Siedepunkt: 50 °C/0,3mmHg Dichte: 1,092 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078660 CAS-Nummer: 62871-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...