2-Chlor-4-hydrazinopyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR60235-250MG
APOSOR60235-250MGEA
29
EUR
APOSOR60235-250MG
2-Chlor-4-hydrazinopyridin
2-Chlor-4-hydrazinopyridin
Formel:
C₅H₆ClN₃ Molecular Weight: 143,58 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460247 CAS-Nummer: 700811-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...