2-Chloro-6-hydrazinopyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-hydrazinylpyrazine

OR60236-1G
APOSOR60236-1GEA 56.7 EUR
APOSOR60236-1G
2-Chloro-6-hydrazinopyrazine
2-Chloro-6-hydrazinopyrazine
Formel: C₄H₅ClN₄
Molecular Weight: 144.56 g/mol
Schmelzpunkt: 136…139 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09049072
CAS-Nummer: 63286-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR