2-Aminopyrimidin-5-carbosäurenitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR60250-1G
APOSOR60250-1GEA
26.1
EUR
APOSOR60250-1G
2-Aminopyrimidin-5-carbosäurenitril 97%
2-Aminopyrimidin-5-carbosäurenitril
Formel:
C₅H₄N₄ Molecular Weight: 120,1121 g/mol Schmelzpunkt: 275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00232791 CAS-Nummer: 1753-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...