2-Bromo-3-methyl-5-nitrobenzyl alcohol ≥95%

Lieferant: Apollo Scientific

OR60271-250MG OR60271-1G
APOSOR60271-250MGEA 241 EUR
APOSOR60271-250MG APOSOR60271-1G
2-Bromo-3-methyl-5-nitrobenzyl alcohol ≥95%
2-Bromo-3-methyl-5-nitrobenzyl alcohol
Formel: C₈H₈BrNO₃
Molecular Weight: 246.05802 g/mol
MDL: MFCD28745354
CAS-Nummer: 1807195-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR