2-Amino-1-(2-naphthyl)-1-ethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Amino-1-hydroxyethyl)naphthalene
OR6056-1G
APOSOR6056-1GEA
369
EUR
APOSOR6056-1G
2-Amino-1-(2-naphthyl)-1-ethanol
2-Amino-1-(2-naphthyl)-1-ethanol
Formel:
C₁₂H₁₃NO Molecular Weight: 187,24 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00974251 CAS-Nummer: 5696-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...