1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
O-(3-Chlorobenzyl)hydroxylamine hydrochloride
OR6062-1G
APOSOR6062-1GEA
205
EUR
APOSOR6062-1G
1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid
1-[(Ammoniooxy)methyl]-3-chlorbenzolchlorid
Formel:
C₇H₉Cl₂NO Schmelzpunkt: 206…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01114569 CAS-Nummer: 29605-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...