2-(4-Methylhomopiperazin-1-yl)benzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: [2-(4-Methylperhydro-1,4-diazepin-1-yl)phenyl]methanol

OR6074-1G
APOSOR6074-1GEA 198 EUR
APOSOR6074-1G
2-(4-Methylhomopiperazin-1-yl)benzyl alcohol 97%
2-(4-Methylhomopiperazin-1-yl)benzyl alcohol
Formel: C₁₃H₂₀N₂O
Molecular Weight: 220,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09065000
CAS-Nummer: 915707-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR