(3-Cyanphenyl)methansulfonsäurechlorid

Lieferant: Apollo Scientific
Gefahr

OR6078-1G
APOSOR6078-1GEA 122 EUR
APOSOR6078-1G
(3-Cyanphenyl)methansulfonsäurechlorid
(3-Cyanophenyl)methanesulphonyl chloride
Formel: C₈H₆ClNO₂S
Molecular Weight: 215,656 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09046692
CAS-Nummer: 56106-01-5
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR