6-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
OR6081-1G
APOSOR6081-1GEA
244
EUR
APOSOR6081-1G
6-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
6-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
Formel:
C₁₀H₁₁N₃ Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09064948 CAS-Nummer: 160017-13-0 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...