2-(4-Methylhomopiperazin-1-yl)benzonitrile

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(2-Cyanophenyl)-4-methyl-1,4-diazepane, 2-(4-Methyl-1,4-diazepan-1-yl)benzonitrile

OR6083-1G
APOSOR6083-1GEA 149 EUR
APOSOR6083-1G
2-(4-Methylhomopiperazin-1-yl)benzonitrile
2-(4-Methylhomopiperazin-1-yl)benzonitrile
Formel: C₁₃H₁₇N₃
Molecular Weight: 215,294 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09065002
CAS-Nummer: 204078-93-3
UN: 2922
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR