Methyl 2-(thiomorpholin-4-yl)isonicotinate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(thiomorpholin-4-yl)pyridine-4-carboxylate

OR6088-1G
APOSOR6088-1GEA 384 EUR
APOSOR6088-1G
Methyl 2-(thiomorpholin-4-yl)isonicotinate 97%
Methyl 2-(thiomorpholin-4-yl)isonicotinate
Formel: C₁₁H₁₄N₂O₂S
Molecular Weight: 238,306 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064957
CAS-Nummer: 898289-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR