Ethyl 3-(1,3-thiazol-2-yl)benzoate 97%

Lieferant: Apollo Scientific
Achtung

OR6089-1G
APOSOR6089-1GEA 149 EUR
APOSOR6089-1G
Ethyl 3-(1,3-thiazol-2-yl)benzoate 97%
Ethyl 3-(1,3-thiazol-2-yl)benzoate
Formel: C₁₂H₁₁NO₂S
Molecular Weight: 233,286 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064945
CAS-Nummer: 886851-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR