4-(Pyridin-3-ylsulphonyl)morpholine 98%

Lieferant: Apollo Scientific

OR6092-1G
APOSOR6092-1GEA 89.9 EUR
APOSOR6092-1G
4-(Pyridin-3-ylsulphonyl)morpholine 98%
4-(Pyridin-3-ylsulphonyl)morpholine
Formel: C₉H₁₂N₂O₃S
Molecular Weight: 228,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090297
CAS-Nummer: 26103-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR