3-[(Pyrrolidin-1-yl)sulphonyl]pyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR6093-1G
APOSOR6093-1GEA
176
EUR
APOSOR6093-1G
3-[(Pyrrolidin-1-yl)sulphonyl]pyridine 98%
3-[(Pyrrolidin-1-yl)sulphonyl]pyridine
Formel:
C₉H₁₂N₂O₂S Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258659 CAS-Nummer: 26103-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...