3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Borono-3-chloro-2-(morpholin-4-yl)pyridine, 4-(4-Borono-3-chloropyridin-2-yl)morpholine
OR6094-100MG
OR6094-50MG
APOSOR6094-100MGEA
132
EUR
APOSOR6094-100MG
APOSOR6094-50MG
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid
Formel:
C₉H₁₂BClN₂O₃ Molecular Weight: 242.467 g/mol Schmelzpunkt: 120…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09258746 CAS-Nummer: 957061-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...