3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Borono-3-chloro-2-(morpholin-4-yl)pyridine, 4-(4-Borono-3-chloropyridin-2-yl)morpholine

OR6094-100MG OR6094-50MG
APOSOR6094-100MGEA 132 EUR
APOSOR6094-100MG APOSOR6094-50MG
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid
Formel: C₉H₁₂BClN₂O₃
Molecular Weight: 242.467 g/mol
Schmelzpunkt: 120…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09258746
CAS-Nummer: 957061-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR