3-(Piperidin-1-ylsulfonyl)pyridin 98%

Lieferant: Apollo Scientific
Achtung

OR6095-1G
APOSOR6095-1GEA 176 EUR
APOSOR6095-1G
3-(Piperidin-1-ylsulfonyl)pyridin 98%
3-(Piperidin-1-ylsulfonyl)pyridin
Formel: C₁₀H₁₄N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD05149384
CAS-Nummer: 26103-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR