6-Methoxyisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
OR61001-5G
OR61001-250MG
OR61001-1G
APOSOR61001-5GEA
0
EUR
APOSOR61001-5G
APOSOR61001-250MG
APOSOR61001-1G
6-Methoxyisatoic anhydride
6-Methoxyisatoic anhydride
Formel:
C₉H₇NO₄ Schmelzpunkt: 258…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11046419 CAS-Nummer: 67765-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...