1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR61002-5G
APOSOR61002-5GEA
132
EUR
APOSOR61002-5G
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester 97%
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester
Formel:
C₁₁H₁₉BN₂O₂ Molecular Weight: 222.0918 g/mol Schmelzpunkt: 107 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664574 CAS-Nummer: 847818-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...