3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride

OR61012-1G OR61012-5G OR61012-10G
APOSOR61012-1GEA 71.3 EUR
APOSOR61012-1G APOSOR61012-5G APOSOR61012-10G
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Formel: C₁₃H₁₆ClNOS
Molecular Weight: 269.79 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011959
CAS-Nummer: 4568-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR