5-Brom-2-cyan-4-methylpyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4-methylpicolinonitrile, 5-Bromo-4-methylpyridine-2-carbonitrile
OR61014-5G
APOSOR61014-5GEA
125
EUR
APOSOR61014-5G
5-Brom-2-cyan-4-methylpyridin 97%
5-Brom-2-cyan-4-methylpyridin
Formel:
C₇H₅BrN₂ Molecular Weight: 197.0300 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07375090 CAS-Nummer: 886364-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...