2-(Aminomethyl)-5-bromo-3-methylpyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-3-methylpyridin-2-yl)methylamine hydrochloride, 5-Bromo-3-methylpicolinylamine hydrochloride, (5-bromo-3-methylpyridin-2-yl)methanamine hydrochloride

OR61016-250MG
APOSOR61016-250MGEA 149 EUR
APOSOR61016-250MG
2-(Aminomethyl)-5-bromo-3-methylpyridine hydrochloride
2-(Aminomethyl)-5-bromo-3-methylpyridine hydrochloride
Formel: C₇H₉BrN₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD17214203
CAS-Nummer: 1257535-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR