2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR61020-1G
APOSOR61020-1GEA 727 EUR
APOSOR61020-1G
2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride
2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride
Formel: C₈H₁₂N₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD17214191
CAS-Nummer: 1257535-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR