6-Chloro-5-methoxynicotinonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR61024-250MG
APOSOR61024-250MGEA
69.6
EUR
APOSOR61024-250MG
6-Chloro-5-methoxynicotinonitrile 97%
6-Chloro-5-methoxynicotinonitrile
Formel:
C₇H₅ClN₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD18071080 CAS-Nummer: 1256835-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...