4-(Methylsulphonyl)benzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

OR61025-1G
APOSOR61025-1GEA 174 EUR
APOSOR61025-1G
4-(Methylsulphonyl)benzene-1,2-diamine
4-(Methylsulphonyl)benzene-1,2-diamine
Formel: C₇H₁₀N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00720917
CAS-Nummer: 21731-57-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR