4-Bromo-5-methyl-1H-indazole, N1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-bromo-5-methyl-1H-indazole-1-carboxylate, 4-Bromo-1-(tert-butoxycarbonyl)-5-methyl-1H-indazole

OR61030-250MG
APOSOR61030-250MGEA 119 EUR
APOSOR61030-250MG
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
4-Bromo-5-methyl-1H-indazole, N1-BOC protected
Formel: C₁₃H₁₅BrN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD16628190
CAS-Nummer: 926922-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR