Methyl 5-bromo-1H-indazole-7-carboxylate

Lieferant: Apollo Scientific
Achtung

OR61032-1G
APOSOR61032-1GEA 108 EUR
APOSOR61032-1G
Methyl 5-bromo-1H-indazole-7-carboxylate
Methyl 5-bromo-1H-indazole-7-carboxylate
Formel: C₉H₇BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08458991
CAS-Nummer: 898747-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR