1H-Indazole-7-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR61033-1G
APOSOR61033-1GEA 400 EUR
APOSOR61033-1G
1H-Indazole-7-carbohydrazide
1H-Indazole-7-carbohydrazide
Formel: C₈H₈N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD11506093
CAS-Nummer: 1086392-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR