3-Amino-5-methylpyridazin 97+%

Lieferant: Apollo Scientific
Achtung

OR61037-1G
APOSOR61037-1GEA 354 EUR
APOSOR61037-1G
3-Amino-5-methylpyridazin 97+%
3-Amino-5-methylpyridazine
Formel: C₅H₇N₃
Molecular Weight: 109,1292 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10698051
CAS-Nummer: 144294-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR