(1-Aminocyclohexyl)methanol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Amino-1-(hydroxyethyl)cyclohexane, 1-Amino-1-cyclohexanemethanol
OR61050-250MG
APOSOR61050-250MGEA
50.9
EUR
APOSOR61050-250MG
(1-Aminocyclohexyl)methanol 98%
(1-Aminocyclohexyl)methanol
Formel:
C₇H₁₅NO Molecular Weight: 129,2 g/mol Schmelzpunkt: 36…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198034 CAS-Nummer: 4313-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...