Piperazine-1-carboxylic acid phenylamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Phenylpiperazine-1-carboxamide

OR61052-500MG
APOSOR61052-500MGEA 145 EUR
APOSOR61052-500MG
Piperazine-1-carboxylic acid phenylamide 95%
Piperazine-1-carboxylic acid phenylamide
Formel: C₁₁H₁₅N₃O
Molecular Weight: 205,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01593572
CAS-Nummer: 115994-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR