6-Bromo-4-nitro-1H-indazole 97%

Lieferant: Apollo Scientific
Achtung

OR61053-5G
APOSOR61053-5GEA 116 EUR
APOSOR61053-5G
6-Bromo-4-nitro-1H-indazole 97%
6-Bromo-4-nitro-1H-indazole
Formel: C₇H₄BrN₃O₂
Molecular Weight: 242.04 g/mol
Schmelzpunkt: 212…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368216
CAS-Nummer: 885518-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR