3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-4-cyclopentylcarbamoylphenylboronic acid, (3-Chloro-4-(cyclopentylcarbamoyl)phenyl)boronic acid

OR6106-1G
APOSOR6106-1GEA 91.9 EUR
APOSOR6106-1G
3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure 98%
3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258664
CAS-Nummer: 957061-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR