2-Bromo-6-iodo-3-methoxypyridine

Lieferant: Apollo Scientific

OR61061-10G OR61061-1G OR61061-5G
APOSOR61061-10GEA 677 EUR
APOSOR61061-10G APOSOR61061-1G APOSOR61061-5G
2-Bromo-6-iodo-3-methoxypyridine
2-Bromo-6-iodo-3-methoxypyridine
Formel: C₆H₅BrINO
Molecular Weight: 313.93 g/mol
Schmelzpunkt: 99…100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09909341
CAS-Nummer: 321535-37-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR