3-Amino-2-methoxypyridine-5-boronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester

OR61065-5G
APOSOR61065-5GEA 66.7 EUR
APOSOR61065-5G
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formel: C₁₂H₁₉BN₂O₃
Molecular Weight: 250.10 g/mol
Schmelzpunkt: 131…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12923427
CAS-Nummer: 893440-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR