2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR61089-250MG
APOSOR61089-250MGEA
127
EUR
APOSOR61089-250MG
2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
2-Bromo-5,5-dimethyl-5,6-dihydrobenzo[d]thiazol-7(4H)-one
Formel:
C₉H₁₀BrNOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD20527744 CAS-Nummer: 10513-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...