5-Bromo-1-(2,6-dichlorophenyl)-7-methoxy-3-[(4-methoxyphenyl)methyl]-4H-quinazolin-2-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR61094-250MG
APOSOR61094-250MGEA
98.6
EUR
APOSOR61094-250MG
5-Bromo-1-(2,6-dichlorophenyl)-7-methoxy-3-[(4-methoxyphenyl)methyl]-4H-quinazolin-2-one 97%
5-Bromo-1-(2,6-dichlorophenyl)-7-methoxy-3-[(4-methoxyphenyl)methyl]-4H-quinazolin-2-one
Formel:
C₂₃H₁₉BrCl₂N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20271802 CAS-Nummer: 444663-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...