3,5-Dibromo-2-hydroxypyrazine

Lieferant: Apollo Scientific
Gefahr

OR61104-1G
APOSOR61104-1GEA 21.9 EUR
APOSOR61104-1G
3,5-Dibromo-2-hydroxypyrazine
3,5-Dibromo-2-hydroxypyrazine
Formel: C4H2Br2N2O
Molecular Weight: 253,8800 g/mol
Schmelzpunkt: 174…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07371406
CAS-Nummer: 21943-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR