2-Chlor-3-hydrazinopyrazin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-hydrazinylpyrazine

OR61106-1G
APOSOR61106-1GEA 60.9 EUR
APOSOR61106-1G
2-Chlor-3-hydrazinopyrazin
2-Chloro-3-hydrazinopyrazine
Formel: C₄H₈Cl₂N₄
Molecular Weight: 144,5600 g/mol
Schmelzpunkt: 154…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08272804
CAS-Nummer: 63286-28-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR