5-Brom-2-cyan-3-methylpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-methylpyridine-2-carbonitrile, 5-Bromo-3-methylpicolinonitrile

OR61109-1G
APOSOR61109-1GEA 21.9 EUR
APOSOR61109-1G
5-Brom-2-cyan-3-methylpyridin
5-Brom-2-cyan-3-methylpyridin
Formel: C₇H₅BrN₂
Molecular Weight: 197,0320 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11975788
CAS-Nummer: 156072-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR