1,3-Benzoxazole-5-boronic acid, pinacol ester 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(1,3-Benzoxazol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole

OR61115-1G
APOSOR61115-1GEA 139 EUR
APOSOR61115-1G
1,3-Benzoxazole-5-boronic acid, pinacol ester 97+%
1,3-Benzoxazole-5-boronic acid, pinacol ester
Formel: C₁₃H₁₆BNO₃
Molecular Weight: 245,082 g/mol
Schmelzpunkt: 88…93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD13181968
CAS-Nummer: 936902-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR