1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
2-Aminopiperidine-4-carboxylic acid, N1-BOC 2-FMOC protected
OR61124-500MG
APOSOR61124-500MGEA
661
EUR
APOSOR61124-500MG
1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid 95+%
1-[(tert-Butoxy)carbonyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-4-caboxylic acid
Formel:
C₂₆H₃₀N₂O₆ Molecular Weight: 466,54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17167329 CAS-Nummer: 1246738-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...