3-Formyl-4-hydroxybenzonitrile

Lieferant: Apollo Scientific

Synonyme: 5-Cyano-2-hydroxybenzaldehyde, 4-Cyano-2-hydroxyphenol

OR61136-1G OR61136-250MG
APOSOR61136-1GEA 169 EUR
APOSOR61136-1G APOSOR61136-250MG
3-Formyl-4-hydroxybenzonitrile
3-Formyl-4-hydroxybenzonitrile
Formel: C₈H₅NO₂
Molecular Weight: 147.131 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09839541
CAS-Nummer: 74901-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR